missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TLR6 Polyclonal Antibody
Beskrivning
TLR6 Polyclonal Antibody for Western Blot, IHC (P), Flow
TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.
Specifikationer
Specifikationer
| Antigen | TLR6 |
| Användningsområden | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.09% sodium azide; pH 7.4 |
| Gen | TLR6 |
| Genaccessionsnr. | Q9EPW9, Q9Y2C9 |
| Gene Alias | CD antigen 286; CD286; TLR6; toll like receptor 6; toll-like receptor 6 |
| Gensymboler | TLR6 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?