Learn More
Brady™ TLS 2200™ and HandiMark™ Label Printers Accessory - Soft Case
Protect your portable label printer and keep accessories organized with this soft carrying case.
Brand: Brady™ 049199
Additional Details : Weight : 0.22000kg
Description
- Package includes the Printer Case only (printer sold separately)
- Designed for use with TLS 2200™ and HandiMark™ Label Printers from Brady™
- This durable and lightweight soft case protects your printer from dust and debris while allowing for easy mobility
- Keeps printer accessories organized and on hand
Specifications
Case | |
Soft Case | |
7 in. |
Black | |
Case | |
177.8 mm |