Läs mer
Tocris Bioscience™ TM 38837
Beskrivning
TM 38837 is a highly potent cannabinoid 1 receptor (CB1) inverse agonist (IC50 values are 8.5 nM and 605 nM for inhibition of [3H]-CP 55940 binding at CB1 and CB2 receptors, respectively). Displays 71-fold selectivity for CB1 receptors over CB2 receptors. Inhibits GTP binding to CB1-overexpressing cell membranes in vitro (EC50 = 18.5 nM). Peripherally restricted. Promotes fear behaviors when given by intracerebrovascular (icv) injection in mice.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-(2,4-Dichlorophenyl)-4-ethyl-N-1-piperidinyl-5-[5-[2-[4-(trifluoromethyl)phenyl]ethynyl]-2-thienyl]-1 H-pyrazole-3-carboxamide |
| CAS | 1253641-65-4 |
| Kvantitet | 5 mg |
| Mål | Cannabinoid R1 / CB1 Inverse Agonists |
| Molekylformel | C30H25Cl2F3N4OS |
| Renhet | 0.98 |