missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM4SF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18239-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
TM4SF5 | |
Polyclonal | |
Unconjugated | |
TM4SF5 | |
TM4SF5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303256, 75604, 9032 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14894 | |
Tm4sf5 | |
TM4SF5 Fusion Protein Ag13004 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |