Läs mer
Invitrogen™ TMBIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551044
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TMBIM1 Polyclonal Antibody detects endogenous levels of TMBIM1 protein.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
Specifikationer
| TMBIM1 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM1 | |
| 2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 316516, 64114, 69660 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BJZ3, Q969X1 | |
| TMBIM1 | |
| Synthesized peptide derived from the C-terminal region of human RECS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |