Läs mer
TMEM119 Antibody (1023426) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) FAB10313B
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifikationer
| TMEM119 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 338773 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 1023426 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| OBIF, Osteoblast Induction Factor, TMEM119 | |
| Chinese Hamster Ovary cell line, CHO derived human TREM-119, Arg26-Met96, Accession # Q4V9L6.1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |