missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Immunogen sequence: RTTAAMWALQ TVEKERGLGG EVPGSHQGPD PYRQLREKDP KYSALRQNFF RYH Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 87%.
Specifikationer
Specifikationer
| Antigen | TMEM205 |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 0.2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | TMEM205 |
| Genaccessionsnr. | Q6UW68 |
| Gene Alias | MBC3205; MGC110858; MGC3205; TMEM205; Transmembrane protein 205; UNQ501; UNQ501/PRO1018 |
| Gensymboler | TMEM205 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?