missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM208 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23882-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
May function as a negative regulator of endoplasmic reticulum-stress induced autophagy.Specifications
TMEM208 | |
Polyclonal | |
Unconjugated | |
TMEM208 | |
HSPC171, TMEM208, transmembrane protein 208 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29100, 66320 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BTX3, Q9CR96 | |
TMEM208 | |
TMEM208 Fusion Protein Ag20845 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |