Learn More
TMEM230 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21466-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Involved in trafficking and recycling of synaptic vesicles.Specifications
TMEM230 | |
Polyclonal | |
Unconjugated | |
TMEM230 | |
C20orf30, HSPC274, TMEM230, UNQ2432/PRO4992 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29058, 681315, 70612 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5BJP5, Q8CIB6, Q96A57 | |
TMEM230 | |
TMEM230 Fusion Protein Ag15080 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |