missing translation for 'onlineSavingsMsg'
Learn More

TMEM61 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20997-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890585

  • 1656.73 SEK / 20µL
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TMEM61
Polyclonal
Unconjugated
TMEM61
TMEM61, transmembrane protein 61
Rabbit
Antigen Affinity Chromatography
RUO
199964
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N0U2
TMEM61
TMEM61 Fusion Protein Ag15044
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers