Learn More
TMEM90A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560041
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ESLSELQNPL LPRSPAHLHG PYPYPETPPS WSCQEKLYSY LLGGAGPAGA HQLLDPGSLQ LAVEAWYRPS CLLGRDKVKE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
TMEM90A Polyclonal antibody specifically detects TMEM90A in Human samples. It is validated for Immunohistochemistry.Specifications
TMEM90A | |
Polyclonal | |
Unconjugated | |
SYNDIG1L | |
CAPUCIN; caudate- and putamen-enriched sequence; caudate-and putamen-enriched sequence; dispanin subfamily C member 1; DSPC1; IFITMD4; interferon induced transmembrane protein domain containing 4; synapse differentiation inducing 1 like; synapse differentiation inducing 1-like; synapse differentiation-inducing gene protein 1-like; SYNDIG1L; TMEM90A; Transmembrane protein 90A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
646658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NDD5 | |
SYNDIG1L | |
Recombinant protein corresponding to Human TMEM90A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only