Learn More
R&D Systems™ TMEM98 Overexpression Lysate
Product Code. 18216170 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. Quantity Unit Size
18216170 0.1 mg 0.1mg
1 options
Product Code. 18216170 Supplier R&D Systems™ Supplier No. NBL117112
 Restricted Item
Beräknad leverans: 07-12-2026
A Novus product, part of R&D Systems.
The TMEM98 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC21829 and based on accession number NM_15544. The protein contains a C-terminal DDK tag.
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens transmembrane protein 98 (TMEM98), transcript variant 1, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Protein State Native
For Use With (Application) Western Blot
For Research Use Only