missing translation for 'onlineSavingsMsg'
Learn More

TMP21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15199-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894274

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer′s disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.
Specifications

Specifications

TMP21
Polyclonal
Unconjugated
TMED10
P24(DELTA), p24delta, S31I125, S31III125, TMED10, Tmp 21 I, TMP21, Transmembrane protein Tmp21
Rabbit
Antigen Affinity Chromatography
RUO
10972, 68581, 84599
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49755, Q63584, Q9D1D4
Tmed10
TMP21 Fusion Protein Ag7347
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions