missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19838-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the disulfide isomerase (PDI) family of endoplasmic reticulum (ER) proteins that catalyze protein folding and thiol-disulfide interchange reactions. The encoded protein has an N-terminal ER-signal sequence, a catalytically active thioredoxin domain, one transmembrane domain and a C-terminal ER-retention sequence. This protein is enriched on the mitochondria-associated-membrane of the ER via palmitoylation of two of its cytosolically exposed cysteines.Specifications
TMX2 | |
Polyclonal | |
Unconjugated | |
TMX2 | |
CGI 31, My009, PDIA12, PIG26, PSEC0045, TMX2, TXNDC14, UNQ237/PRO270 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295701, 51075, 66958 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XIK2, Q9D710, Q9Y320 | |
TMX2 | |
TMX2 Fusion Protein Ag13828 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |