Läs mer
PeproTech TNF alpha Polyclonal Antibody, PeproTech®, Invitrogen™
Beskrivning
AA Sequence of recombinant protein: VRSSSRTPSD KPVAHVVANP QAEGQLQWLN RRANALLANG VELRDNQLVV PSEGLYLIYS QVLFKGQGCP STHVLLTHTI SRIAVSYQTK VNLLSAIKSP CQRETPEGAE AKPWYEPIYL GGVFQLEKGD RLSAEINRPD YLDFAESGQV YFGIIAL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human TNF-alpha. Anti-Human TNF-alpha-specific antibody was purified by affinity chromatography employing an immobilized Human TNF-alpha matrix. Sandwich ELISA: To detect Human TNF-alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TNF-alpha (500-P31AGBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TNF-alpha. Western Blot: To detect Human TNF-alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human TNF-alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifikationer
Specifikationer
| Antigen | TNF alpha |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.1-1.0 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with no preservative |
| Gen | Tnf |
| Genaccessionsnr. | P01375 |
| Gene Alias | APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF α; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFα; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor |
| Gensymboler | Tnf |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.