missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MedChemExpress TNF-alpha/TNFSF2 Protein, Human
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Specifications
Specifications
| Innehåll och lagring | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Kvantitet | 100 μg |
| Frakt skick | Room temperature |
| Molekylvikt (g/mol) | Approximately 17.3 kDa |
| Synonym | rHuTNF-α/TNFSF2∣ TNF-alpha∣ Cachectin∣ DIF∣ TNFA∣ Differentiation-inducing factor |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction