missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TNF Mouse anti-Human, BV421, Clone: MAb11, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566275
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TNF Monoclonal antibody specifically detects TNF in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifikationer
| TNF | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| MAb11 | |
| Vol. Per test: 5μL | |
| Tumor necrosis factor alpha; TNF-a; TNF-α; TNFSF2; Cachectin | |
| Recombinant Human TNF | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG1 κ |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Reseach Use Only.
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering