missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFR2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19272-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.Specifications
TNFR2 | |
Polyclonal | |
Unconjugated | |
TNFRSF1B | |
CD120b, Etanercept, p75, p75TNFR, p80 TNF alpha receptor, TBP 2, TBPII, TNF R II, TNF R2, TNF R75, TNF Receptor 2, TNF Receptor II, TNF RII, TNFBR, TNFR II, TNFR1B, TNFR2, TNFR80, TNFRSF1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21938, 7133 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20333, P25119 | |
TNFRSF1B | |
TNFR2 Fusion Protein Ag5866 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |