missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF13C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22582-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13.1-q13.31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.Specifications
TNFRSF13C | |
Polyclonal | |
Unconjugated | |
TNFRSF13C | |
BAFF R, BAFF receptor, BAFFR, BLyS receptor 3, BR3, CD268, TNFRSF13C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115650 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96RJ3 | |
TNFRSF13C | |
TNFRSF13C Fusion Protein Ag18202 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |