missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TNFRSF17 (Human) Recombinant Protein
Human TNFRSF17 full-length ORF ( AAH58291, 1 a.a. - 184 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000608-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLQMAGQCSQNEYFDSLLHACIPCQLRCSSNTPPLTCQRYCNASVTNSVKGTNAILWTCLGLSLIISLAVFVLMFLLRKISSEPLEDEFKNTGSGLLGMANIDLEKSRTGDEIILPRGLEYTVEECTCEDCIKSKPKVDSDHCFPLPAMEEGATILVTTKTNDYCKSLPAALSATEIEKSISAR
Specifications
AAH58291 | |
tumor necrosis factor receptor superfamily, member 17 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TNFRSF17 | |
GST |
608 | |
Wheat germ expression system | |
10 μg | |
BCM, BCMA, CD269 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |