missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11862-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes all the forms of TNFSF13B such as soluble form (17 kDa), membrane form (32-35 kDa) and glycosylation form (45-52 kDa).
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells.Specifications
TNFSF13B | |
Polyclonal | |
Unconjugated | |
TNFSF13B | |
B cell activating factor, B lymphocyte stimulator, BAFF, BLYS, CD257, DTL, TALL 1, TALL1, THANK, TNFSF13B, TNFSF20, ZTNF4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10673, 24099 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9WU72, Q9Y275 | |
TNFSF13B | |
TNFSF13B Fusion Protein Ag2440 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |