missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TNPO1 (Human) Recombinant Protein
Human TNPO1 partial ORF ( NP_002261, 25 a.a. - 141 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_002261 |
---|---|
Gene ID (Entrez) | 3842 |
Name | transportin 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177181
|
Abnova™
H00003842-Q01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16187181
|
Abnova™
H00003842-Q01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: PDTTIQRTVQQKLEQLNQYPDFNNYLIFVLTKLKSEDEPTRSLSGLILKNNVKAHFQNFPNGVTDFIKSECLNNIGDSSPLIRATVGILITTIASKGELQNWPDLLPKLCSLLDSED
Specifications
NP_002261 | |
transportin 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
IPO2, KPNB2, MIP, MIP1, TRN | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3842 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TNPO1 | |
GST |