missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11580-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.Specifications
TNS4 | |
Polyclonal | |
Unconjugated | |
TNS4 | |
C terminal tensin like protein, CTEN, PP14434, tensin 4, TNS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217169, 303517, 84951 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4V8I3, Q8BZ33, Q8IZW8 | |
TNS4 | |
TNS4 Fusion Protein Ag2169 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |