missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TNS4 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunofluorescence, Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878753
|
Proteintech
11580-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868753
|
Proteintech
11580-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.Specifications
TNS4 | |
Immunofluorescence, Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q4V8I3, Q8BZ33, Q8IZW8 | |
217169, 303517, 84951 | |
TNS4 Fusion Protein Ag2169 | |
Primary | |
-20°C | |
TNS4 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C terminal tensin like protein, CTEN, PP14434, tensin 4, TNS4 | |
TNS4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |