missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tobramycin, MP Biomedicals™
1305.00 SEK - 6155.00 SEK
Chemical Identifiers
CAS | 32986-56-4 |
---|---|
Molecular Formula | C18H37N5O9 |
Molecular Weight (g/mol) | 467.52 |
InChI Key | NLVFBUXFDBBNBW-PBSUHMDJSA-N |
Synonym | Distobram, Gernebcin |
PubChem CID | 36294 |
ChEBI | CHEBI:28864 |
IUPAC Name | (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol |
SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495321
|
MP Biomedicals
0215692690 |
500mg |
6155.00 SEK
500mg |
Estimated Shipment: 27-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11485321
|
MP Biomedicals
0215692680 |
0.1g |
1305.00 SEK
100mg |
Estimated Shipment: 27-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
32986-56-4 | |
467.52 | |
Distobram, Gernebcin | |
CHEBI:28864 | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N |
C18H37N5O9 | |
NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
36294 | |
(2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol |
Specifications
Tobramycin | |
C18H37N5O9 | |
NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
(2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol | |
36294 | |
467.5 | |
White |
32986-56-4 | |
Distobram, Gernebcin | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N | |
467.52 | |
CHEBI:28864 | |
Powder | |
500mg |