Läs mer
Tocainide Hydrochloride, LoGiCal
Produktkod. 30459684
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30459684 5 mg
1 options
Produktkod. 30459684 Leverantör LoGiCal Leverantörsnummer LGCFOR3323.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tocainide Hydrochloride
CAS 71395-14-7
Analyt- eller komponentnamn Tocainide Hydrochloride
Frakt skick Room Temperature
Molekylformel C11 H16 N2 O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Tocainide Hydrochloride, Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, hydrochloride (1:1), Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, monohydrochloride, Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, monohydrochloride, (±)-, 2-Amino-2',6'-propionoxylidide hydrochloride, Taquidil, Tocainide-hydrochloride, Tonocard, Xylotocan
InChI formel InChI=1S/C11H16N2O.ClH/c1-7-5-4-6-8(2)10(7)13-11(14)9(3)12;/h4-6,9H,12H2,1-3H3,(H,13,14);1H
Lösningstyp Neat
LEDER Cl.CC(N)C(=O)Nc1c(C)cccc1C
IUPAC-namn 2-amino-N-(2,6-dimethylphenyl)propanamide;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 228.72
Kvantitet 5 mg
Formel vikt 228.1029 g/mol