missing translation for 'onlineSavingsMsg'
Learn More

Tocainide Hydrochloride, TRC

Product Code. 30452187
Change view
Click to view available options
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30452187 100 mg
30450898 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452187 Supplier Toronto Research Chemicals Supplier No. TRCT525450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71395-14-7
Molekylformel C11 H16 N2 O . Cl H
Molekylvikt (g/mol) 228.72
Synonym Tocainide Hydrochloride, Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, hydrochloride (1:1), Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, monohydrochloride, Propanamide, 2-amino-N-(2,6-dimethylphenyl)-, monohydrochloride, (±)-, 2-Amino-2',6'-propionoxylidide hydrochloride, Taquidil, Tocainide-hydrochloride, Tonocard, Xylotocan
IUPAC-namn 2-amino-N-(2,6-dimethylphenyl)propanamide;hydrochloride
LEDER Cl.CC(N)C(=O)Nc1c(C)cccc1C

Specifications

Kemiskt namn eller material Tocainide Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C11H16N2O.ClH/c1-7-5-4-6-8(2)10(7)13-11(14)9(3)12;/h4-6,9H,12H2,1-3H3,(H,13,14);1H
Formel vikt 228.1029
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.