Läs mer
Tocopherols (Technical Grade), TRC
Produktkod. 30449937
Change view
Kvantitet:
100 g
Produktkod. Kvantitet
30449937 100 g
1 options
Produktkod. 30449937 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT730500100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tocopherols
CAS 1406-66-2
Molekylformel C29 H50 O2
Kvantitet 100 g
Synonym AT 160, Controx VP, Copherol F 1300, Covi-Ox T 50, Decanox MTS 50, Dry E-Mix 20, E-Mix 35L, ’ E-Oil 800, E-Oil Super 60, Fortium MTD 10, Golden Tocopherol 70, Irganox E 217DF, Riken Dry E-Mix SP-T, Riken E Oil Super 80G, Riken E-Emulsion, Tocoblend L 50IP, Tocomix L 70, Tocopherol 80, Tocovet E 1000, Tokomikkusu 500
InChI formel InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21?,22?,29-/m0/s1
LEDER CC(C)CCCC(C)CCCC(C)CCC[C@@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1
IUPAC-namn (2S)-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol
Molekylvikt (g/mol) 430.706
Formel vikt 430.381