Läs mer
R&D Systems™ GLYX 13
Beskrivning
GLYX 13 is a NMDA receptor partial agonist that acts at the glycine site. Simultaneously acts as a promoter of the induction of long-term potentiation (LTP) and as a suppressor of long-term depression (LTD). Also promotes VGF and BDNF release and increases ERK/mTOR signaling . Exhibits nootropic, neuroprotective and antinociceptive activity, and enhances learning, memory and cognition in vivo. Brain penetrant.
Specifikationer
Specifikationer
| Produktlinje | Tocris |
| Renhet | 0.95 |
| Kvantitet | 1 mg |