missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ TLQP 21
GREENER_CHOICE

Produktkod. 18044642
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18044642 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18044642 Leverantör Tocris Bioscience™ Leverantörsnummer 3051/1

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

VGF-derived peptide

TLQP 21 is a VGF-derived peptide; spans residues 556-576 of the precursor sequence. Protects cerebellar granule cells (CGCs) from serum and potassium deprivation-induced apoptosis. Increases energy expenditure and prevents early phase diet-induced diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Renhet 95%
Innehåll och lagring Store at -20°C
Kvantitet 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.