Läs mer
Tofacitinib impurity L, TRC
Produktkod. 30454376
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30454376 2.5 mg 2.5mg
30450435 10 mg -
30450841 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5280152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1675248-19-7
Molekylformel C16H22N6O2
Molekylvikt (g/mol) 330.38
Synonym (3R,4R)-4-Methyl-3-(methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-β-oxo-1-piperidinepropanamide, Tofacitinib Citrate Impurity 05
IUPAC-namn 3-[(3R,4R)-4-methyl-3-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]piperidin-1-yl]-3-oxopropanamide
LEDER C[C@@H]1CCN(C[C@@H]1N(C)c2ncnc3[nH]ccc23)C(=O)CC(=O)N
Specifikationer
Kemiskt namn eller material Tofacitinib impurity L
Kvantitet 2.5 mg
InChI formel InChI=1S/C16H22N6O2/c1-10-4-6-22(14(24)7-13(17)23)8-12(10)21(2)16-11-3-5-18-15(11)19-9-20-16/h3,5,9-10,12H,4,6-8H2,1-2H3,(H2,17,23)(H,18,19,20)/t10-,12+/m1/s1
Formel vikt 330.1804