Läs mer
Tofacitinib Impurity P, TRC
Produktkod. 30453139
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453139 10 mg
30456494 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30453139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT52802510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1252883-90-1
Molekylformel C20H25N5
Molekylvikt (g/mol) 335.45
Synonym N-Methyl-N-[(3S,4S)-4-methyl-1-(phenylmethyl)-3-piperidinyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine, N-((3S,4S)-1-Benzyl-4-methylpiperidin-3-yl)-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
IUPAC-namn N-[(3S,4S)-1-benzyl-4-methylpiperidin-3-yl]-N-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
LEDER C[C@H]1CCN(Cc2ccccc2)C[C@H]1N(C)c3ncnc4[nH]ccc34
Specifikationer
Kemiskt namn eller material Tofacitinib Impurity P
Kvantitet 10 mg
InChI formel InChI=1S/C20H25N5/c1-15-9-11-25(12-16-6-4-3-5-7-16)13-18(15)24(2)20-17-8-10-21-19(17)22-14-23-20/h3-8,10,14-15,18H,9,11-13H2,1-2H3,(H,21,22,23)/t15-,18+/m0/s1
Formel vikt 335.211