missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tofisopam, LoGiCal
Specifications
Specifications
| Kemiskt namn eller material | Tofisopam |
| CAS | 22345-47-7 |
| Analyt- eller komponentnamn | Tofisopam |
| Frakt skick | Room Temperature |
| Molekylformel | C22 H26 N2 O4 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 5H-2,3-Benzodiazepine, 1-(3,4-dimethoxyphenyl)-5-ethyl-7,8-dimethoxy-4-methyl-1-(3,4-Dimethoxyphenyl)-4-methyl-5-ethyl-7,8-dimethoxy-5H-2,3-benzodiazepine, 7,8-Dimethoxy-1-(3,4-dimethoxyphenyl)-5-ethyl-4-methyl-5H-2,3-benzodiazepine, EGYT 341, Grandaxin, Seriel, Tofisopam |
| InChI formel | InChI=1S/C22H26N2O4/c1-7-15-13(2)23-24-22(14-8-9-18(25-3)19(10-14)26-4)17-12-21(28-6)20(27-5)11-16(15)17/h8-12,15H,7H2,1-6H3 |
| Lösningstyp | Neat |
| LEDER | CCC1C(=NN=C(c2ccc(OC)c(OC)c2)c3cc(OC)c(OC)cc13)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?