Läs mer
Tolafentrine, TRC
Produktkod. 30453844
Change view
Kvantitet:
0.5 mg
2 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30453844 2 mg -
30450736 0.5 mg 0.5mg
30449071 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453844 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5310002MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139308-65-9
Molekylformel C28 H31 N3 O4 S
Molekylvikt (g/mol) 505.63
IUPAC-namn N-[4-[(4aS,10bR)-8,9-dimethoxy-2-methyl-3,4,4a,10b-tetrahydro-1H-benzo[c][1,6]naphthyridin-6-yl]phenyl]-4-methylbenzenesulfonamide
LEDER COc1cc2[C@@H]3CN(C)CC[C@@H]3N=C(c4ccc(NS(=O)(=O)c5ccc(C)cc5)cc4)c2cc1OC
Specifikationer
Kemiskt namn eller material Tolafentrine
Kvantitet 2 mg
InChI formel InChI=1S/C28H31N3O4S/c1-18-5-11-21(12-6-18)36(32,33)30-20-9-7-19(8-10-20)28-23-16-27(35-4)26(34-3)15-22(23)24-17-31(2)14-13-25(24)29-28/h5-12,15-16,24-25,30H,13-14,17H2,1-4H3/t24-,25-/m0/s1
Formel vikt 505.2035