Learn More
Tolazoline, TRC
Product Code. 30455882
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
Product Code. Quantity Unit Size
30455882 2 g 2g
1 options
Product Code. 30455882 Supplier Toronto Research Chemicals Supplier No. TRCT5350032G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tolazoline
CAS 59-98-3
Molecular Formula C10H12N2
Quantity 2 g
Synonym 2-Imidazoline','2-benzyl- (8CI), Imidazoline','2-benzyl- (6CI), 4,5-Dihydro-2-(phenylmethyl)-1H-imidazole, 2-Benzyl-2-imidazoline, 2-Benzyl-4,5-dihydro-1H-imidazole, 2-Benzyl-4,5-imidazoline, 2-Benzylimidazoline, Artonil, Benzazoline, Benzidazol, Benzylimidazoline, Dilatol ASI, Divascol, Imidalin, Imidaline, Kasimid, Lambril, NSC 35110, Olitensol, Peripherine, Phenylmethylimidazoline, Prefaxil, Priscol, Priscoline, Vasimid, Vasodil
InChI Formula InChI=1S/C10H12N2/c1-2-4-9(5-3-1)8-10-11-6-7-12-10/h1-5H,6-8H2,(H,11,12)
SMILES C(C1=NCCN1)c2ccccc2
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol) 160.22
Formula Weight 160.1