Läs mer
Tolcide 2230 (90%), TRC
Produktkod. 30011466
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011466 100 mg
30004565 250 mg
30005982 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT535280100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21564-17-0
Molekylformel C9 H6 N2 S3
Molekylvikt (g/mol) 238.35
Synonym TCMTB (2-(Thiocyanomethylthio)benzothiazole), Thiocyanic acid, (2-benzothiazolylthio)methyl ester, (2-Benzothiazolylthio)methyl thiocyanate, 2-(Thiocyanatomethylthio)-1,3-benzothiazole, 2-(Thiocyanatomethylthio)benzo[d]thiazole, 2-(Thiocyanomethylthio)benzothiazole, 2-(Thiocyanomethylthio)benzthiazole, 2-[(Thiocyanatomethyl)thio]benzothiazole, Acticide WB 300, Afrotin CRO, Alentisan, Argent, Argent 30, Ascend, BN 30, Benthiazole, Bulab 6009, Busan 1030, Busan 1118, Busan 15, Busan 30, Busan 30-1, Busan 30A, Busan 30I, Busan 30L, Busan 30WB, Busan 30WBA, Busan 70, Busan 71, Busan 72, Busan 72A, Busan 80, Delsan 30, Fungicide FDE, Guzafan, Ichiban, KVK 733059, Nu-flow T, Nusan, Sancelant TMB, Superdavloxan, TCMTB, Tolcide 2230
IUPAC-namn 1,3-benzothiazol-2-ylsulfanylmethyl thiocyanate
LEDER N#CSCSc1nc2ccccc2s1
Specifikationer
Kemiskt namn eller material TCMTB (Busan)
Kvantitet 100 mg
InChI formel InChI=1S/C9H6N2S3/c10-5-12-6-13-9-11-7-3-1-2-4-8(7)14-9/h1-4H,6H2
Formel vikt 237.9693