Läs mer
Tolclofos-methyl-oxon, Dr. Ehrenstorfer
Produktkod. 30321021
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30321021 10 mg
1 options
Produktkod. 30321021 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREC17590200
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tolclofos-methyl-oxon
CAS 97483-08-4
Analyt- eller komponentnamn Tolclofos-methyl-oxon
Frakt skick +20°C
Molekylformel C9 H11 Cl2 O4 P
Certifikat och efterlevnad ISO/IEC 17025
Synonym Tolclofos-methyl Oxon, Phosphoric acid, 2,6-dichloro-4-methylphenyl dimethyl ester, Tolclofos-methyl-oxon;
InChI formel InChI=1S/C9H11Cl2O4P/c1-6-4-7(10)9(8(11)5-6)15-16(12,13-2)14-3/h4-5H,1-3H3
LEDER COP(=O)(OC)Oc1c(Cl)cc(C)cc1Cl
IUPAC-namn (2,6-dichloro-4-methylphenyl) dimethyl phosphate
Molekylvikt (g/mol) 285.061
Kvantitet 10 mg
Formel vikt 283.977 g/mol