Läs mer
Tolpronine Hydrochloride, LoGiCal
Produktkod. 30462900
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462900 10 mg
1 options
Produktkod. 30462900 Leverantör LoGiCal Leverantörsnummer LGCFOR3271.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tolpronine Hydrochloride
CAS 6775-25-3
Analyt- eller komponentnamn Tolpronine Hydrochloride
Frakt skick Room Temperature
Molekylformel C15 H21 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Tolpronine hydrochloride, 1(2H)-Pyridineethanol, 3,6-dihydro-alpha-[(2-methylphenoxy)methyl]-, hydrochloride (1:1), 1(2H)-Pyridineethanol, 3,6-dihydro-alpha-[(2-methylphenoxy)methyl]-, hydrochloride (9CI), 1(2H)-Pyridineethanol, 3,6-dihydro-alpha-[(o-tolyloxy)methyl]-, hydrochloride (8CI)
InChI formel InChI=1S/C15H21NO2.ClH/c1-13-7-3-4-8-15(13)18-12-14(17)11-16-9-5-2-6-10-16;/h2-5,7-8,14,17H,6,9-12H2,1H3;1H
Lösningstyp Neat
LEDER Cl.Cc1ccccc1OCC(O)CN2CCC=CC2
IUPAC-namn 1-(3,6-dihydro-2H-pyridin-1-yl)-3-(2-methylphenoxy)propan-2-ol;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 283.79
Kvantitet 10 mg
Formel vikt 283.1339 g/mol