missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tolrestat, MedChemExpress
Description
Tolrestat is a potent, orally active aldose reductase inhibitor with IC50 of 35 nM.
Specifications
Specifications
| Kemiskt namn eller material | Tolrestat |
| CAS | 82964-04-3 |
| Färg | Light Yellow |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C16H14F3NO3S |
| Kvantitet | 5 mg |
| Synonym | AY-27773 |
| Löslighetsinformation | DMSO : 50 mg/mL (139.92 mM; Need ultrasonic) |
| LEDER | O=C(O)CN(C(C1=C2C=CC(OC)=C(C(F)(F)F)C2=CC=C1)=S)C |
| Molekylvikt (g/mol) | 357.35 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?