missing translation for 'onlineSavingsMsg'
Learn More

Toluene, BAKER ANALYZED HPLC, J.T. Baker™

Catalog Number p-7138052
Click to view available options
Quantity:
1 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

Get rapid, reproducible performance and separation results. Toluene, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use
Melting Point -95°C
Absorbance Max. 0.40 at 288nm, Max. 0.10 at 300nm, Max. 0.01 at 350nm
Color Colorless
Linear Formula C6H5CH3
Solubility Information 0.5g/L in water
Formula Weight 92.14g/mol
Percent Purity ≥99.7%
Grade For use in High Performance Liquid Chromatography
Density 0.87/cm3
Boiling Point 110°C to 111°C
Quantity 1 L
Explosion Limit 1.2 - 7 Vol %
Flash Point 4°C
Ignition Point 535°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 3 ppm
Viscosity 0.6 mPas at 20°C
Water Max. 0.02%
Chemical Name or Material Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'