Learn More
Tolyltriazole, TRC
Specifikationer
Specifikationer
| Chemical Name or Material | Tolyltriazole |
| CAS | 29385-43-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 2 C7 H7 N3 |
| Quantity | 5 g |
| Synonym | 1H-Benzotriazole','6(or 7)-methyl-, 1H-Benzotriazole','4(or 5)-methyl- (9CI), 6(or 7)-Methyl-1H-benzotriazole, Additin RC 8221, C.V.I. TT, Cobratec TT 100, Cobratec TT 35I, Iramet 39, Methylbenzotriazole, Preventol CI 7-100, Seetec T, Seetec TT, Seetec TT-R, Stabinol MBTZ, TTZ, Tolyltriazole |
| InChI Formula | InChI=1S/2C7H7N3/c1-5-2-3-6-7(4-5)9-10-8-6;1-5-3-2-4-6-7(5)9-10-8-6/h2*2-4H,1H3,(H,8,9,10) |
| SMILES | Cc1ccc2[nH]nnc2c1.Cc3cccc4[nH]nnc34 |
| IUPAC Name | 4-methyl-1H-benzotriazole;5-methyl-1H-benzotriazole |
| Molecular Weight (g/mol) | 266.3 |
| Visa mer |