Läs mer
Tomatidine Hydrochloride ( >85%), TRC
Produktkod. 30449622
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30449622 50 mg
1 options
Produktkod. 30449622 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT53850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tomatidine hydrochloride
CAS 6192-62-7
Molekylformel C27 H45 NO2 . x HCl
Kvantitet 50 mg
Synonym (3beta,5alpha,22beta,25S)-Spirosolan-3-ol Hydrochloride, Tomatidin Hydrochloride
InChI formel InChI=1S/C27H45NO2.ClH/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4;/h16-24,28-29H,5-15H2,1-4H3;1H/t16-,17-,18-,19-,20+,21-,22-,23-,24-,25-,26-,27+;/m0./s1
LEDER O[C@H]1CC[C@@]2(C)[C@](C1)(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])C[C@@]5([H])[C@]4([H])[C@H](C)[C@@]6(NC[C@@H](C)CC6)O5)[H].[xHCl]
Molekylvikt (g/mol) 452.11
Formel vikt 451.3217