Läs mer
Tomatine (85%), TRC
Produktkod. 30011021
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011021 1 mg
30005841 10 mg
30009312 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5385101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17406-45-0
Molekylformel C50 H83 N O21
Molekylvikt (g/mol) 1034.19
Synonym Tomatine (8CI), α-Tomatine (6CI,7CI), (3β,5α,22β,25S)-Spirosolan-3-yl O-β-D-glucopyranosyl-(1→2)-O-[β-D-xylopyranosyl-(1→3)]-O-β-D-glucopyranosyl-(1→4)-β-D-galactopyranoside, Spiro[8H-naphth[2',1':4,5]indeno[2,1-b]furan-8,2'-piperidine], β-D-galactopyranoside deriv., Spirosolane, β-D-galactopyranoside deriv., Lycopersicin, NSC 234440, NSC 9223
LEDER C[C@H]1CCC2(NC1)O[C@H]3C[C@H]4[C@@H]5CC[C@H]6C[C@H](CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C)O[C@@H]7O[C@H](CO)[C@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O[C@@H]9OC[C@@H](O)[C@H](O)[C@H]9O)[C@H]8O[C@@H]%10O[C@H](CO)[C@@H](O)[C@H](O)[C@H]%10O)[C@H](O)[C@H]7O
Specifikationer
Kemiskt namn eller material Lycopersicin (Tomatine)
Kvantitet 1 mg
InChI formel InChI=1S/C50H83NO21/c1-20-7-12-50(51-15-20)21(2)32-28(72-50)14-26-24-6-5-22-13-23(8-10-48(22,3)25(24)9-11-49(26,32)4)65-45-40(63)37(60)41(31(18-54)68-45)69-47-43(71-46-39(62)36(59)34(57)29(16-52)66-46)42(35(58)30(17-53)67-47)70-44-38(61)33(56)27(55)19-64-44/h20-47,51-63H,5-19H2,1-4H3/t20-,21-,22-,23-,24+,25-,26-,27+,28-,29+,30+,31+,32-,33-,34+,35+,36-,37+,38+,39+,40+,41-,42-,43+,44-,45+,46-,47-,48-,49-,50?/m0/s1
Formel vikt 1033.5458