missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16689-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TOMM6, also named as OBTP and TOM6, is a part of the preprotein translocase complex of the outer mitochondrial membrane (TOM complex).Specifications
TOMM6 | |
Polyclonal | |
Unconjugated | |
TOMM6 | |
OBTP, TOM6, TOMM6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100188893, 66119, 681123 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96B49, Q9CQN3 | |
Tomm6 | |
TOMM6 Fusion Protein Ag10037 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |