Läs mer
Tocris Bioscience™ Tomoxetine hydrochloride
GREENER_CHOICE
Produktkod. 18005522
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18005522 10 mg
18045982 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18005522 Leverantör Tocris Bioscience™ Leverantörsnummer 2011/10
Potent and selective noradrenalin re-uptake inhibitor
Tomoxetine hydrochloride is a potent and selective noradrenalin re-uptake inhibitor (Ki values are 5, 77 and 1451 nM for inhibition of radioligand binding to human NET, SERT and DAT respectively). Displays minimal affinity for a range of other neurotransmitter receptors and transporters (Ki > 1 μM). Antidepressant.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82248-59-7
Molekylformel C17H22ClNO
Specifikationer
Kemiskt namn eller material Tomoxetine hydrochloride
Kvantitet 10 mg
Formel vikt 291.82
Renhet >99%
Molekylvikt (g/mol) 291.819