Learn More
Uvex™ Top Grade 7200 Welding Gloves
Catalog Number 15357032
Click to view available options
10
Robust split cowhide leather welding gloves are sewn with aramid fiber thread for increased protection and heat resistance.
  • Cowhide leather welding gloves sewn with aramid fiber threads
  • Gloves provide mechanical abrasion resistance, tear propagation properties, and heat resistance
  • Extended gauntlet cuff offers forearm protection
Color Black
Material Split Cowhide Leather, Aramid Fiber
Size 10
EN388 Standard 4244X
Glove Cuff Style Gauntlet
Series Top Grade 7200
Length (Metric) 35 cm