Learn More
Abnova™ TOP2A/CEN17q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0007.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16012985
  • 15410.00 SEK / 100µL
Estimated Shipment: 10-07-2023
to see stock.
Includes: DAPI Counterstain (1500ng/mL) 125μL
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
DAPI Counterstain (1500 ng/mL ) 250 uL
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
100 μL
Texas Red, FITC
Store at 4°C in the dark.
TOP2A/CEN17q FISH Probe
TOP2A, CEN17q
17q21, 17q11.2