Läs mer
TopFluor Cholesterol, TRC
Produktkod. 30455925
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30455925 2.5 mg 2.5mg
30455774 1 mg -
30449181 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455925 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5403302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 878557-19-8
Molekylformel C36H51BF2N2O
Molekylvikt (g/mol) 576.61
Synonym (T-4)-[(3beta)-24-(3,5-dimethyl-1H-pyrrol-2-yl-kN)-24-(3,5-dimethyl-2H-pyrrol-2-ylidene-kN)chol-5-en-3-olato]difluoroboron, Bodipy FL Cholesterol, Bodipy cholesterol
LEDER [F-][B+3]1([F-])[N]2=C(C)C=C(C)C2=C(CC[C@@H](C)[C@H]3CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C)C7=C(C)C=C(C)[N-]17
Specifikationer
Kemiskt namn eller material TopFluor Cholesterol
Kvantitet 2.5 mg
InChI formel InChI=1S/C36H51BF2N2O/c1-21(8-10-29-33-22(2)18-24(4)40(33)37(38,39)41-25(5)19-23(3)34(29)41)30-12-13-31-28-11-9-26-20-27(42)14-16-35(26,6)32(28)15-17-36(30,31)7/h9,18-19,21,27-28,30-32,42H,8,10-17,20H2,1-7H3/t21-,27+,28+,30-,31+,32+,35+,36-/m1/s1
Formel vikt 576.4063