Learn More
Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with MA Circulators
For working temperatures from -50deg.C to +200°C. Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with MA Circulators features increased functionality and additional warning and safety functions.
Brand: Julabo™ 9153625
Additional Details : Weight : 0.50000kg
Description
- PID2 temperature control, stability ±0.02°C
- ATC 3-point calibration
- RS232 interface
- Early warning system for low liquid level and high/low temperature
- Pump capacity electronically adjustable
Specifications
Interior (L x W x D): 14 x 12 x 14cm | |
Classification III (FL) according to DIN 12876-1 | |
-28°C to +200°C | |
2 Barbed fittings for tubing 8 and 10mm I.D. (pump connections M10 x 1 female) | |
F25-MA | |
42 x 23 x 61 cm |
4.5 L | |
0.26kW at +20°C, 0.2kW at 0°C, 0.06kW at -20°C | |
2 kW | |
Stainless Steel | |
±0.02°C | |
115 V 60 Hz 13 A |