missing translation for 'onlineSavingsMsg'
Learn More

TOR1AIP2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24769-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16873255

  • 4721.67 SEK / 150µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.
Specifications

Specifications

TOR1AIP2
Polyclonal
Unconjugated
TOR1AIP2
LULL1, Lumenal domain like LAP1, RP11 12M5.5, TOR1AIP2, torsin A interacting protein 2
Rabbit
Antigen Affinity Chromatography
RUO
163590
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H496
TOR1AIP2
TOR1AIP2 Fusion Protein Ag20126
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers